SY3H3249F5F8 Synthonix Corporation

3-Difluoromethylpiperidine hydrochloride

  • MDL number MFCD18325187
  • Cas Number : 1093759-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6079.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6079.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart