SY3H3249F601 Synthonix Corporation

3,3-Difluorocyclopentaecarboxylic acid

  • MDL number MFCD15526661
  • Cas Number : 1260897-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8067.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8067.html
hstatements H302 , H315 , H317 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart