SY3H3249F603 Synthonix Corporation

(4,4-Dimethyloxetan-2-yl)methylamine

  • MDL number MFCD22628734
  • Cas Number : 1408075-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/8432.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8432.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart