SY3H3249F609 Synthonix Corporation

Ethyl 5-oxothiepane-4-carboxylate

  • MDL number MFCD25562903
  • Cas Number : 925218-54-8
  • Molecular Weight : 202.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/10055.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/10055.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart