SY3H3249F611 Synthonix Corporation

Ethyl 1-Boc-3-fluoropiperidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1228631-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11477.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11477.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart