SY3H3249F613 Synthonix Corporation

1-Boc-3-ethynyl-3-hydroxypyrrolidine

  • MDL number No MDL found
  • Cas Number : 275387-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11721.html

Safety Information

MSDS https://synthonix.com/11721.html
hstatements H302 ,
Availability details for
Quantity
  • Availability
  • Add to Cart