SY3H3249F614 Synthonix Corporation

Ethyl 1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate

  • MDL number MFCD16036533
  • Cas Number : 1067915-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1234.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1234.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart