SY3H3249F634 Synthonix Corporation

Ethyl 2-Cyano-4,4-diethoxybutyrate

  • MDL number No MDL found
  • Cas Number : 52133-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15568.html

Safety Information

MSDS https://synthonix.com/15568.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart