SY3H3249F645 Synthonix Corporation

ethyl 3-oxocyclohexane-1-carboxylate

  • MDL number MFCD00205583
  • Cas Number : 33668-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/23119.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23119.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart