SY3H3249F6A4 Synthonix Corporation

Ethyl 1-benzyl-4-oxopyrrolidine-3-carboxylate

  • MDL number MFCD09998956
  • Cas Number : 1027-35-6
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6950.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/6950.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart