SY3H3249F6A7 Synthonix Corporation

Ethyl 2-bromo-2-cyclopropylacetate

  • MDL number MFCD18837719
  • Cas Number : 1200828-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7311.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7311.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart