SY3H3249F6A9 Synthonix Corporation

Ethyl 1-Boc-3-pyrrolidine acetate

  • MDL number MFCD11977331
  • Cas Number : 664364-29-8
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8027.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/8027.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart