SY3H3249F6AD Synthonix Corporation

1-(5-fluoropyridin-2-yl)ethanamine

  • MDL number MFCD18260255
  • Cas Number : 915720-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0635.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0635.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart