SY3H3249F6B2 Synthonix Corporation

3-Fluoro-4-piperidinone hydrochloride

  • MDL number No MDL found
  • Cas Number : 1070896-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11694.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11694.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart