SY3H3249F6B3 Synthonix Corporation

3-Fluorotetrahydropyran-4,4-diol

  • MDL number No MDL found
  • Cas Number : 1523571-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/12100.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/12100.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart