SY3H3249F6C4 Synthonix Corporation

2-Formyl-1-trityl-aziridine

  • MDL number MFCD20926171
  • Cas Number : 173277-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1841.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1841.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart