SY3H3249F6EE Synthonix Corporation

4-Fluoro-1H-benzo[d]imidazol-2(3H)-one

  • MDL number No MDL found
  • Cas Number : 256519-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/38220.html

Safety Information

MSDS https://synthonix.com/38220.html
Availability details for
Quantity
  • Availability
  • Add to Cart