SY3H3249F72E Synthonix Corporation

3-Fluorodihydro-2H-pyran-4(3H)-one

  • MDL number MFCD11977351
  • Cas Number : 624734-19-6
  • Molecular Weight : 118.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5739.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5739.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart