SY3H3249F738 Synthonix Corporation

3-Hydroxy-1-methylpiperidine

  • MDL number MFCD00006495
  • Cas Number : 3554-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0286.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0286.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart