SY3H3249F739 Synthonix Corporation

4-Hydroxymethyl-1-methyl-2-piperidone

  • MDL number MFCD17015932
  • Cas Number : 20845-32-3
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0356.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0356.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart