SY3H3249F757 Synthonix Corporation

1-Boc-3-hydroxy-4-methylenepiperidine

  • MDL number MFCD15474939
  • Cas Number : 159635-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14885.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/14885.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart