SY3H3249F76F Synthonix Corporation

5-Hydroxy-4-methylfuran-2(5H)-one

  • Cas Number : 40834-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32216.html

Safety Information

MSDS https://synthonix.com/32216.html
Availability details for
Quantity
  • Availability
  • Add to Cart