SY3H3249F79E Synthonix Corporation

5-(Hydroxymethyl)dihydrofuran-2(3H)-one

  • MDL number MFCD11706998
  • Cas Number : 10374-51-3
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5013.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5013.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart