SY3H3249F7A0 Synthonix Corporation

3-Hydroxy-1-oxa-8-azaspiro[4.5]decane-8-carboxylic acid tert-butyl ester

  • MDL number MFCD09749842
  • Cas Number : 240401-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form White to Pale-yellow Solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/6090.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/6090.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart