SY3H3249F7A2 Synthonix Corporation

2-Boc-5-hydroxy-2-azaspiro[3.3]heptane

  • MDL number MFCD20230648
  • Cas Number : 1330764-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7218.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7218.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart