SY3H3249F7A3 Synthonix Corporation

2-Boc-6-hydroxy-2-azaspiro[3.4]octane

  • MDL number MFCD22415223
  • Cas Number : 1363381-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7221.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7221.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart