SY3H3249F7A6 Synthonix Corporation

6-hydroxy-3-Azabicyclo[3.1.1]heptane-3-carboxylic acid 1,1-dimethylethyl ester

  • MDL number MFCD17168421
  • Cas Number : 1357353-36-6
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7839.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/7839.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart