SY3H3249F7A8 Synthonix Corporation

3-(Hydroxymethyl)-3-methylpiperidine hydrochloride

  • MDL number MFCD22421713
  • Cas Number : 955027-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7938.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7938.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart