SY3H3249F7EF Synthonix Corporation

(3-Methylpyrrolidin-3-yl)methanamine dihydrochloride

  • MDL number MFCD19443964
  • Cas Number : 1630907-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/1124.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1124.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart