SY3H3249F7F6 Synthonix Corporation

2-Methyl-1,3,7-triazaspiro[4.5]dec-2-en-4-one hydrochloride

  • MDL number MFCD11869758
  • Cas Number : 1214028-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37406.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37406.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart