SY3H3249F801 Synthonix Corporation

Methyl aminotetrahydropyran-4-ylacetate

  • MDL number MFCD03425227
  • Cas Number : 477585-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0708.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0708.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart