SY3H3249F804 Synthonix Corporation

Methyl 1-Cbz-piperidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 174543-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/10282.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10282.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart