SY3H3249F819 Synthonix Corporation

1-Boc-3-methylpyrrolidine-3-methanol

  • MDL number No MDL found
  • Cas Number : 1263506-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11405.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11405.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart