SY3H3249F81F Synthonix Corporation

3-Methoxypiperidine

  • MDL number MFCD05022463
  • Cas Number : 4045-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1167.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1167.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart