SY3H3249F84B Synthonix Corporation

1-(1-Methyl-1H-pyrazol-4-yl)ethylamine

  • MDL number No MDL found
  • Cas Number : 911788-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/14047.html

Safety Information

MSDS https://synthonix.com/14047.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart