SY3H3249F856 Synthonix Corporation

Methyl 3,3-difluorocyclopentanecarboxylate

  • MDL number No MDL found
  • Cas Number : 1394129-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/14942.html

Safety Information

MSDS https://synthonix.com/14942.html
hstatements H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart