SY3H3249F865 Synthonix Corporation

4-methoxyazepane

  • MDL number No MDL found
  • Cas Number : 1071594-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16379.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/16379.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart