SY3H3249F877 Synthonix Corporation

4-Methoxy-hexahydro-1H-azepine hydrochloride

  • MDL number MFCD21641983
  • Cas Number : 1408076-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1861.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1861.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart