SY3H3249F8AE Synthonix Corporation

methyl 2,2-dicyanocyclopropane-1-carboxylate

  • MDL number MFCD29060186
  • Cas Number : 39822-30-5
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/29834.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/29834.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart