SY3H3249F8B0 Synthonix Corporation

Methyl 1,4'-bipiperidine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 889952-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/29983.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/29983.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart