SY3H3249F8C0 Synthonix Corporation

trans-Methyl 4-methylpyrrolidine-3-carboxylate

  • MDL number MFCD11845468
  • Cas Number : 1065331-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32340.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32340.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart