SY3H3249F8C2 Synthonix Corporation

Methyl 2-amino-3,3,3-trifluoropropanoate hydrochloride

  • MDL number MFCD08532471
  • Cas Number : 134297-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32350.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32350.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart