SY3H3249F9AB Synthonix Corporation

Methyl 5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD08275678
  • Cas Number : 35309-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/5245.html

Safety Information

MSDS https://synthonix.com/5245.html
Availability details for
Quantity
  • Availability
  • Add to Cart