SY3H3249F9AE Synthonix Corporation

methyl 2-amino-2-(oxetan-3-yl)acetate hemioxalate

  • MDL number MFCD26959140

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6273.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6273.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart