SY3H3249F9AF Synthonix Corporation

3-Methyloxetan-2-one

  • MDL number MFCD22055146
  • Cas Number : 1823-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/6342.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6342.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart