SY3H3249F9B3 Synthonix Corporation

6-Methyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine

  • MDL number MFCD00805140
  • Cas Number : 7496-50-6
  • Molecular Weight : 168.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7020.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7020.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart