SY3H3249F9B7 Synthonix Corporation

1-Methylpyrrolidine-3-ethanol

  • MDL number MFCD17015913
  • Cas Number : 36763-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7517.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7517.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart