SY3H3249F9BA Synthonix Corporation

Methyl 1-methyl-2-oxo-4-piperidinecarboxylate

  • MDL number MFCD13705362
  • Cas Number : 20845-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7546.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7546.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart