SY3H3249F9BD Synthonix Corporation

Methyl 2-oxopiperidine-4-carboxylate

  • MDL number MFCD17012745
  • Cas Number : 25504-47-6
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7686.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7686.html
pictograms GHS05, GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart