SY3H3249F9BE Synthonix Corporation

Methyl 6-bromo-2-oxo-1,2,3,4-tetrahydro-1,8-naphthyridine-3-carboxylate

  • MDL number MFCD08275095
  • Cas Number : 335031-10-2
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7745.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7745.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart